5-Amino-2-methylbenzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-3-(trifluoromethyl)aniline

001827-25G 001827-5G
101775-556EA 193.85 USD
101775-556 101775-554
5-Amino-2-methylbenzotrifluoride ≥97%
5-Amino-2-methylbenzotrifluoride

Hazard: Irritant-Harmful

Formula: C₈H₈F₃N
MW: 175.16 g/mol
Boiling Pt: 85-86°C/8mm
Density: 1.24
MDL Number: MFCD01631582
CAS Number: 65934-74-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 85-86°/8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR