6-Methoxy-m-toluenesulfonyl chloride
Supplier: Matrix Scientific
001786-1G
001786-5G
101775-548EA
16.92
USD
101775-548
101775-550
6-Methoxy-m-toluenesulfonyl chloride
6-Methoxy-m-toluenesulfonyl chloride
Hazard: Corrosive/Moisture Sensitive/Refrigerate
Formula:
C₈H₉ClO₃S Melting Pt: 82…84 °C |
MDL Number:
MFCD00060552 CAS Number: 88040-86-2 |
Specification Test Results
Melting point | 82-84° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...