1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2'-(Benzene-1,3-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

001769-100G 001769-5G 001769-25G
101775-544EA 761.54 USD
101775-544 101775-540 101775-542
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥97%
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene

TCA listed: T
Hazard: Corrosive/Irritant

Formula: C₁₂H₆F₁₂O₂
MW: 410.16
Boiling Pt: 99…100 °C (20 mmHg)
MDL Number: MFCD00042090
CAS Number: 802-93-7

Order Now

Specification Test Results

Boiling point 99-100°/20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR