4-Fluoro-2-(trifluoromethyl)aniline ≥97%

Supplier: Matrix Scientific

Synonyms: α,α,α,4-Tetrafluoro-o-toluidine, 2-Amino-alpha,alpha,alpha,5-tetrafluorotoluene, 2-Amino-5-fluorobenzotrifluoride, 2-Amino-α,α,α,5-tetrafluorotoluene

001731-25G 001731-100G
101775-526EA 35.38 USD
101775-526 101775-528
4-Fluoro-2-(trifluoromethyl)aniline ≥97%
4-Fluoro-2-(trifluoromethyl)aniline

Hazard: Toxic

Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 70
Density: 1.38
MDL Number: MFCD00007831
CAS Number: 393-39-5
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 70-72°/17.
Density 1.38
ND20 1.464


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR