4-Bromo-2-fluorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

001711-25G 001711-5G
101775-520EA 87.69 USD
101775-520 101775-518
4-Bromo-2-fluorobenzenesulfonyl chloride ≥98%
4-Bromo-2-fluorobenzenesulphonyl chloride

Hazard: Corrosive

Formula: C₆H₃BrClFO₂S
MW: 273.51 g/mol
Boiling Pt: 84 °C (0.5 mmHg)
Melting Pt: 63…66 °C
MDL Number: MFCD01090993
CAS Number: 216159-03-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 84°/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR