α,4-Dibromo-2-fluorotoluene ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-1-(bromomethyl)-2-fluorobenzene, 4-Bromo-2-fluorobenzyl bromide
001707-25G
001707-100G
101775-514EA
23.08
USD
101775-514
101775-516
α,4-Dibromo-2-fluorotoluene ≥97%
α,4-Dibromo-2-fluorotoluene
Hazard: Corrosive/Lachrymator
Formula:
C₇H₅Br₂F MW: 267.92 g/mol Melting Pt: 30°C |
MDL Number:
MFCD00055467 CAS Number: 76283-09-5 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 30° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...