2-Bromo-4-fluoroaniline ≥97%

Supplier: Matrix Scientific

001687-25G 001687-100G
101775-506EA 18.46 USD
101775-506 101775-508
2-Bromo-4-fluoroaniline ≥97%
2-Bromo-4-fluoroaniline

Hazard: Toxic/Irritant

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 221 °C
Density: 1.67
MDL Number: MFCD00042462
CAS Number: 1003-98-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 221°
Density 1.67
ND20 1.583

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR