4-Bromo-3-fluoroaniline ≥98%

Supplier: Matrix Scientific

001686-25G 001686-100G
101775-502EA 83.08 USD
101775-502 101775-504
4-Bromo-3-fluoroaniline ≥98%
4-Bromo-3-fluoroaniline

Hazard: Toxic/Irritant-Harmful

Formula: C₆H₅BrFN
Melting Pt: 72…73 °C
MDL Number: MFCD00672933
CAS Number: 656-65-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 72-73°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR