Ethyl-3,4-difluorobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3,4-difluorobenzoate

001668-25G 001668-100G
101775-496EA 50.77 USD
101775-496 101775-498
Ethyl-3,4-difluorobenzoate ≥97%
Ethyl-3,4-difluorobenzoate

Hazard: Flammable

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
Boiling Pt: 96°C/15mm
MDL Number: MFCD00272138
CAS Number: 144267-96-9

Order Now

Specification Test Results

Boiling point 96°/15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR