Ethyl-2,4-difluorobenzoate ≥97%
Supplier: Matrix Scientific
001667-5G
001667-25G
101775-490EA
27.69
USD
101775-490
101775-492
Ethyl-2,4-difluorobenzoate ≥97%
Hazard: Flammable
Formula:
C₉H₈F₂O₂ Boiling Pt: 65…67 °C (15 mmHg) |
MDL Number:
MFCD00153149 CAS Number: 108928-00-3 |
Specification Test Results
Boiling point | 65-67°/15m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...