Ethyl-4,4-difluoroacetoacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-3-oxobutyrate

001666-25G 001666-100G
101775-486EA 58.46 USD
101775-486 101775-488
Ethyl-4,4-difluoroacetoacetate ≥95%
Ethyl-4,4-difluoroacetoacetate

TCA listed: T
Hazard: Flammable/Irritant

Formula: C₆H₈F₂O₃
Boiling Pt: 162°C
MDL Number: MFCD00039277
CAS Number: 352-24-9

Order Now

Specification Test Results

Boiling point 162°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR