4-Chloro-3-(trifluoromethyl)aniline ≥99%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-chloro-α,α,α-trifluorotoluene, 1-Amino-4-chloro-3-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride

001632-100G 001632-25G
101775-476EA 60 USD
101775-476 101775-474
4-Chloro-3-(trifluoromethyl)aniline ≥99%
4-Chloro-3-(trifluoromethyl)aniline

TCA listed: T
Hazard: Toxic

Formula: C₇H₅ClF₃N
Boiling Pt: 132 °C (27 Torr)
Melting Pt: 36…38 °C
MDL Number: MFCD00007846
CAS Number: 320-51-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 36-38°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR