2-Bromo-2-butene cis- and trans-mixture ≥95%

Supplier: Matrix Scientific

001598-25G 001598-100G
101775-464EA 130.77 USD
101775-464 101775-466
2-Bromo-2-butene cis- and trans-mixture ≥95%
2-Bromo-2-butene

TCA listed: T
Hazard: Flammable

Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 82…90 °C
Density: 1.328
Flash Pt: 1 °C
MDL Number: MFCD00000141
CAS Number: 13294-71-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 82-90°
Density 1.328
ND20 1.4613

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR