4,4,5,5,5-Pentafluoropentan-1-ol ≥93%

Supplier: Matrix Scientific

Synonyms: 4,4,5,5,5-Pentafluoropentanol

001568-5G 001568-25G
101775-454EA 43.08 USD
101775-454 101775-456
4,4,5,5,5-Pentafluoropentan-1-ol ≥93%
4,4,5,5,5-Pentafluoropentan-1-ol

Hazard: Flammable

Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 62-64°C/35mm
Density: 1.35
MDL Number: MFCD00153224
CAS Number: 148043-73-6

Order Now

Specification Test Results

Boiling point 62-64°/35m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR