2,3,4,5,6-Pentafluorotoluene ≥99%

Supplier: Matrix Scientific

001556-25G 001556-100G
101775-432EA 153.85 USD
101775-432 101775-434
2,3,4,5,6-Pentafluorotoluene ≥99%
2,3,4,5,6-Pentafluorotoluene

TCA listed: T
Hazard: Flammable

Formula: C₇H₃F₅
MW: 182.09 g/mol
Boiling Pt: 117 °C
Density: 1.439
Flash Pt: 34 °C
MDL Number: MFCD00000298
CAS Number: 771-56-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 117°
Density 1.439
ND20 1.403

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR