2,2,3,3,3-Pentafluoropropyl trifluoromethanesulfonate ≥97%
Supplier: Matrix Scientific
001552-1G
001552-5G
101775-426EA
86.15
USD
101775-426
101775-428
2,2,3,3,3-Pentafluoropropyl trifluoromethanesulfonate ≥97%
2,2,3,3,3-Pentafluoropropyl trifluoromethanesulfonate
Hazard: Toxic/Corrosive
Formula:
C₄H₂F₈O₃S MW: 282.11 g/mol Boiling Pt: 102-105°C/740mm |
MDL Number:
MFCD01862006 CAS Number: 6401-00-9 |
Specification Test Results
Boiling point | 102-105°/7 |
ND20 | 1.3032 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...