2-Hydroxy-5-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)pyridin-2-ol

001502-5G 001502-25G
101775-410EA 27.69 USD
101775-410 101775-412
2-Hydroxy-5-(trifluoromethyl)pyridine ≥98%
2-Hydroxy-5-(trifluoromethyl)pyridine

Hazard: Corrosive/Irritant

Formula: C₆H₄F₃NO
MW: 163.1 g/mol
Melting Pt: 145…148 °C
MDL Number: MFCD00042315
CAS Number: 33252-63-0

Order Now

Specification Test Results

Melting point 145-148°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR