2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97%

Supplier: Matrix Scientific

001500-1G 001500-5G
101775-398EA 43.08 USD
101775-398 101775-400
2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97%
2'-Chloro-5'-(trifluoromethyl)acetophenone

Hazard: Flammable

Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
Boiling Pt: 98-100°C/6mm
Density: 1.392
Flash Pt: 91°C
MDL Number: MFCD00045092
CAS Number: 71648-45-8

Order Now

Specification Test Results

Boiling point 98-100°/6m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR