2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Nitro-5-(trifluoromethyl)pyridin-2(1H)-one

001464-5G 001464-1G
101775-396EA 95.38 USD
101775-396 101775-394
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine ≥98%
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine

Hazard: Toxic/Stored under Argon

Formula: C₆H₃F₃N₂O₃
MW: 208.096 g/mol
Melting Pt: 189…190 °C
MDL Number: MFCD00153193
CAS Number: 33252-64-1

Order Now

Specification Test Results

Melting point 183-186°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR