4-Methoxy-3-(trifluoromethyl)benzoyl chloride

Supplier: Matrix Scientific

001448-1G 001448-5G
101775-390EA 30.77 USD
101775-390 101775-392
4-Methoxy-3-(trifluoromethyl)benzoyl chloride
4-Methoxy-3-(trifluoromethyl)benzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₉H₆ClF₃O₂
MW: 238.59
Boiling Pt: 244
Melting Pt: 75
MDL Number: MFCD01631576
CAS Number: 98187-18-9
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR