4-Methoxy-3-(trifluoromethyl)benzonitrile

Supplier: Matrix Scientific

001447-1G 001447-5G
101775-386EA 30.77 USD
101775-386 101775-388
4-Methoxy-3-(trifluoromethyl)benzonitrile
4-Methoxy-3-(trifluoromethyl)benzonitrile

Hazard: Toxic

Formula: C₉H₆F₃NO
MW: 201.15 g/mol
Melting Pt: 98-100°C
MDL Number: MFCD01631575
CAS Number: 261951-87-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 98-100°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR