4-Bromo-2-(trifluoromethoxy)aniline ≥98%

Supplier: Matrix Scientific

001385-100G 001385-5G 001385-25G
101775-374EA 136.92 USD
101775-374 101775-370 101775-372
4-Bromo-2-(trifluoromethoxy)aniline ≥98%
4-Bromo-2-(trifluoromethoxy)aniline

Hazard: Toxic/Irritant

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
Boiling Pt: 98…100 °C (15 mmHg)
Density: 1.691
MDL Number: MFCD00179338
CAS Number: 175278-09-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 98-100°/15

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR