4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-(trifluoromethoxy)benzene-1-sulphonyl chloride

001383-1G 001383-25G 001383-5G
101775-362EA 29.23 USD
101775-362 101775-366 101775-364
4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride ≥97%
4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride

Hazard: Corrosive/Refrigerate

Formula: C₇H₃BrClF₃O₃S
MW: 339.52 g/mol
Boiling Pt: 86…88 °C (0.2 mmHg)
MDL Number: MFCD00179336
CAS Number: 175278-14-5

Order Now

Specification Test Results

Boiling point 86-88°/0.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR