2-Nitro-4-(trifluoromethyl)phenylacetonitrile

Supplier: Matrix Scientific

Synonyms: 2-(2-Nitro-4-(trifluoromethyl)phenyl)acetonitrile

001355-1G 001355-5G
101775-356EA 43.08 USD
101775-356 101775-358
2-Nitro-4-(trifluoromethyl)phenylacetonitrile
2-Nitro-4-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅F₃N₂O₂
Melting Pt: 36…38 °C
MDL Number: MFCD00045621
CAS Number: 13544-06-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 36-38°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR