2-Amino-6-(trifluoromethoxy)benzothiazole ≥97%

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole

001333-25G 001333-5G
101775-348EA 176.92 USD
101775-348 101775-346
2-Amino-6-(trifluoromethoxy)benzothiazole ≥97%
2-Amino-6-(trifluoromethoxy)benzothiazole

Hazard: Toxic

Formula: C₈H₅F₃N₂OS
Melting Pt: 119 °C
MDL Number: MFCD00210213
CAS Number: 1744-22-5

Order Now

Specification Test Results

Melting point 119°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR