4,4,5,5,6,6,6-Heptafluorohexanoic acid ≥97%

Supplier: Matrix Scientific

001296-1G 001296-5G 001296-25G
101775-334EA 146.15 USD
101775-334 101775-336 101775-338
4,4,5,5,6,6,6-Heptafluorohexanoic acid ≥97%
4,4,5,5,6,6,6-Heptafluorohexanoic acid

Hazard: Corrosive

Formula: C₆H₅F₇O₂
Melting Pt: 38…39 °C
MDL Number: MFCD00077553
CAS Number: 356-02-5

Order Now

Specification Test Results

Melting point 38-39°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR