1-Bromo-3,5-difluorobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
3,5-Difluorobromobenzene
001287-100G
001287-500G
101775-328EA
44.62
USD
101775-328
101775-330
1-Bromo-3,5-difluorobenzene ≥98%
1-Bromo-3,5-difluorobenzene
Hazard: Flammable
Formula:
C₆H₃BrF₂ MW: 192.99 g/mol Boiling Pt: 140°C Density: 1.676 |
MDL Number:
MFCD00010305 CAS Number: 461-96-1 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 140° |
Density | 1.676 |
ND20 | 1.499 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...