Dextran sulfate sodium salt, Powder

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dextran sulfate sodium from DextranDextran sulphate sodium from Dextran

DE133-5GM DE133-25GM DE133-100GM
700013-146EA 953.7 USD
700013-146 700013-148 700013-150
Dextran sulfate sodium salt, Powder
Dextran sulphate sodium salt

Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 12,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.

Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR