Cocarboxylase ≥97%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Thiamine pyrophosphate chloride, Aneurinepyrophosphoric acid, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy(phosphonooxy)phosphoryl)oxy)ethyl)-4-methylthiazol-3-ium chloride, Cocarboxylase chloride
T1586-5GM
T1586-100GM
T1586-25GM
T1586-5KGBL
700011-508EA
271.08
USD
700011-508
700011-504
700011-506
700011-510
Cocarboxylase ≥97%
Cocarboxylase
Thiamine Pyrophosphate Chloride, also known as Thiamine diphosphate, works as a coenzyme in many enzymatic reactions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₂H₁₉ClN₄O₇P₂S MW: 460.77 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00038740 CAS Number: 154-87-0 EINECS: 205-836-1 |
Specification Test Results
Assay (%) | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...