Orotic acid, anhydrous
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
O2142-50GM
O2142-25KGBL
O2142-250GM
700009-454EA
175
USD
700009-454
700009-452
700009-450
Orotic acid, anhydrous
Orotic acid
Orotic Acid, Anhydrous, also known as pyrimidinecarboxylic acid, is a heterocyclic compound and an acid used as a mineral carrier in several dietary supplements like lithium orotate.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₅H₄N₂O₄ MW: 156.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006027 CAS Number: 65-86-1 EINECS: 200-619-8 |
Specification Test Results
Assay (%) | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...