Acriflavine hydrochloride
Supplier: SPECTRUM CHEMICAL MFG CORP
AC175-5GM
AC175-100GM
AC175-5KGBL
AC175-25GM
700009-236EA
72.5
USD
700009-236
700009-232
700009-238
700009-234
Acriflavine hydrochloride
Acriflavine hydrochloride
Acriflavine Hydrochloride, a topical antiseptic, is a dye and can be used to treat fungal infections.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₂₇H₂₈Cl₄N₆ MW: 578.36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069039 CAS Number: 8063-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...