Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt

AD125-100GM AD125-5KGBL
700009-070EA 158.8 USD
700009-070 700009-066 700009-072
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥99%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt)

Adenosine-5'-triphosphate Disodium Salt is a nucleoside triphosphate that cells use as a coenzyme. It is used by structural proteins and enzymes in several types of cellular processes that include motility and biosynthetic reactions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃
MW: 551.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00080339
CAS Number: 987-65-5
EINECS: 213-579-1

Order Now

Specification Test Results

Assay (%) 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR