(2-Hydroxylpropyl)-γ-cyclodextrine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2-Hydroxypropyl)-γ-Cyclodextrin, HGC

H2758-100GM H2758-5GM
700008-512EA 667.75 USD
700008-512 700008-514
(2-Hydroxylpropyl)-γ-cyclodextrine
(2-Hydroxylpropyl)-γ-cyclodextrine

(2-Hydroxypropyl)-gamma-cyclodextrin is a hydrophilic compound that can be used as a complexing agent to increase the aqueous solubility and dissolution rate.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇₂H₁₂₈O₄₈
MW: 1761.7600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148890
CAS Number: 128446-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR