Creatine (N-amidinosarcosine), anhydrous ≥98% suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine

C1522-100GM C1522-25GM
700007-922EA 161.81 USD
700007-922 700007-924
Creatine (N-amidinosarcosine), anhydrous ≥98% suitable for use as excipient
Creatine (N-amidinosarcosine)
Formula: C₄H₉N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00004282
CAS Number: 57-00-1
EINECS: 200-306-6

Order Now

Specification Test Results

Assay (%) 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR