(2-Hydroxylpropyl)-β-cyclodextrine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

H2690-10KGBL H2690-25GM H2690-5GM H2690-100GM
700007-876EA 8666.39 USD
700007-876 700007-878 700007-880 700007-874
(2-Hydroxylpropyl)-β-cyclodextrine
(2-Hydroxylpropyl)-β-cyclodextrine

(2-Hydroxypropyl)-beta-cyclodextrin is cyclic polysaccharide comprised of 7 glucopyranose units. This ingredient appears as a solid and functions as a chelating agent. Cyclodextrins are able to form host-guest complexes with hydrophobic molecules and are used in the production cholesterol free products. Beta-cyclodextrin complexes with certain carotenoid food colorants have been shown to intensify color, increase water solubility and improve light stability

Formula: C₆₃H₁₁₂O₄₂
Storage Temperature: Ambient
MDL Number: MFCD16621721
CAS Number: 128446-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR