Glycylglycine (Gly-Gly)

Supplier: SPECTRUM CHEMICAL MFG CORP

G1024-25GM G1024-250GM G1024-100GM
700007-696EA 110.32 USD
700007-696 700007-694 700007-692
Glycylglycine (Gly-Gly)
Proteins and Peptides

Glycylglycine, Ultrapure, also known as Diglycine, is used in the synthesis of complex peptides. Ultrapure products supplied by Spectrum are indicative of a grade suitable for research purposes and typically are not suitable for human consumption or therapeutic use.


  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning
  • Use for Trace analysis

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR