L(-)-Carnitine free base ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

C1166-5KGBL C1166-25GM C1166-500GM C1166-50KGBL C1166-100GM
700007-578EA 4395.85 USD
700007-578 700007-572 700007-574 700007-576 700007-570
L(-)-Carnitine free base ≥97%
L(-)-Carnitine

L-Carnitine, Free Base is a quaternary ammonium compound and most commonly used as a nutritional supplement.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038747
CAS Number: 541-15-1
EINECS: 208-768-0

Order Now

Specification Test Results

Assay (%) 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR