Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥98%
Supplier: Spectrum Chemical Mfg. Corp.
Ethylene Glycol Dimethacrylate is a diester that is formed by the condensation of ethylene glycol and methacrylic acid and used primarily in free radical copolymer crosslinking reactions. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₁₀H₁₄O₄ Density: 1,051 Storage Temperature: Refrigerator |
MDL Number:
MFCD00008590 CAS Number: 97-90-5 EINECS: 202-617-2 |
Specification Test Results
Assay (%) | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...