Dipropyleneglycol (mixture of isomers) ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dipropylene glycol
D1207-1LT
D1207-20LT
D1207-4LTGL
D1207-200LTB
700003-498EA
215.39
USD
700003-498
700003-502
700003-506
700003-500
Dipropyleneglycol (mixture of isomers) ≥99%
Dipropyleneglycol (mixture of isomers)
Dipropylene Glycol, also known as 1,1'-Oxybis(2-propanol), is primarily used as a plasticizer, as a solvent, and as a polymerization initiator or monomer.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₁₄O₃ MW: 134.18 g/mol Density: 1.023 Storage Temperature: Ambient |
MDL Number:
MFCD00051023 CAS Number: 25265-71-8 EINECS: 246-770-3 |
Specification Test Results
Assay (%) | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...