Dipropyleneglycol monomethyl ether (mixture of isomers) ≥99%

Supplier: Spectrum Chemical Mfg. Corp.

D1110-25ML D1110-1LT D1110-20LT
700003-494EA 79.85 USD
700003-494 700003-490 700003-492
Dipropyleneglycol monomethyl ether (mixture of isomers) ≥99%

Dipropylene Glycol Methyl Ether is a colorless liquid with a mild and pleasant odor. It is a solvent used in paints, pastes, dyes, resins, brake fluids, inks and in making cosmetics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₇H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00059604
CAS Number: 34590-94-8
EINECS: 252-104-2

Order Now

Specification Test Results

Assay (%) 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR