L(-)-Sorbose ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one
, L-(-)-Sorbose
S0066-25G
TCS0066-025GEA
108.19
USD
TCS0066-025G
L(-)-Sorbose ≥98.0% (by HPLC)
L(-)-Sorbose
CAS Number: 87-79-6
MDL Number: MFCD00151097
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 165
Specific rotation [a]20/D: -43 deg (C=1, H2O)
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 380 °C (1013 hPa) Melting Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151097 CAS Number: 87-79-6 EINECS: 201-771-8 Merck Index: 12,08874 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...