(+)-5,6-O-Isopropylidene-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (R)-5-((S)-2,2-Dimethyl-1,3-dioxolan-4-yl)-3,4-dihydroxyfuran-2(5H)-one

I0507-5G I0507-25G
TCI0507-005GEA 146 USD
TCI0507-005G TCI0507-25G
(+)-5,6-O-Isopropylidene-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)
(+)-5,6-O-Isopropylidene-L-ascorbic acid

CAS Number: 15042-01-0
MDL Number: MFCD00010552
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210
Specific rotation [a]20/D: 10 deg (C=5, MeOH)

Formula: C₉H₁₂O₆
MW: 216.19 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00010552
CAS Number: 15042-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f992aa101d701e6765e75f1c257508dddc4697f7&vid=f992aa101d701e6765e75f1c257508dddc4697f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR