N-Chlorosuccinimide ≥98.0% (by titrimetric analysis) for synthesis
Supplier: TCI America
Synonyms:
1-Chloropyrrolidine-2,5-dione
C0291-500G
C0291-25G
TCC0291-500GEA
69
USD
TCC0291-500G
TCC0291-025G
N-Chlorosuccinimide ≥98.0% (by titrimetric analysis) for synthesis
N-Chlorosuccinimide
CAS Number: 128-09-6
Molecular Formula: C4H4ClNO2
Molecular Weight: 133.53
Form: Crystals
Color: White
Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction
Formula:
C₄H₄ClNO₂ MW: 133.53 g/mol |
MDL Number:
MFCD00005511 CAS Number: 128-09-6 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Melting point (°C) | 148 |
Storage Temperature | 0-10°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...