ω-Bromoacetophenone ≥98.0%

Supplier: TCI America
Warning

Synonyms: α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone

B0535-25G B0535-100G
TCB0535-025GEA 62.8 USD
TCB0535-025G TCB0535-500G TCB0535-100G
ω-Bromoacetophenone ≥98.0%
ω-Bromoacetophenone

CAS Number: 70-11-1
MDL Number: MFCD00000195
Molecular Formula: C8H7BrO
Molecular Weight: 199.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 50
Flash Point (°C): 113
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Bromoacetophenone

Formula: (C₆H₅)C(O)CH₂Br
MW: 199.05 g/mol
Boiling Pt: 135 °C
Melting Pt: 50 °C
Density: 1.647 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000195
CAS Number: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Merck Index: 13,01386
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bbe81b3062537887d6fe4bd1987fd4983720c70&vid=8bbe81b3062537887d6fe4bd1987fd4983720c70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR