Triphenyl borate ≥96.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Phenyl Borate, Boric Acid Triphenyl Ester
B0524-25G
B0524-500G
TCB0524-025GEA
86
USD
TCB0524-025G
TCB0524-500G
Triphenyl borate ≥96.0% (by titrimetric analysis)
Triphenyl borate
CAS Number: 1095-03-0
MDL Number: MFCD00059011
Molecular Formula: C18H15BO3
Molecular Weight: 290.13
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 101
Other Product Names / Synonyms: Boric Acid Triphenyl Ester, Phenyl Borate
Formula:
C₁₈H₁₅BO₃ MW: 290.13 g/mol Boiling Pt: 158 °C Melting Pt: 101 °C |
MDL Number:
MFCD00059011 CAS Number: 1095-03-0 |
Specification Test Results
Purity/Analysis Method | >96.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...