4-Bromoantipyrine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one

B0515-5G B0515-25G
TCB0515-005GEA 58 USD
TCB0515-005G TCB0515-025G
4-Bromoantipyrine ≥98.0% (by HPLC, titration analysis)
4-Bromoantipyrine

CAS Number: 5426-65-3
MDL Number: MFCD00020779
Molecular Formula: C11H11BrN2O
Molecular Weight: 267.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 117

Formula: C₁₁H₁₁BrN₂O
MW: 267.13 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00020779
CAS Number: 5426-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2b49480ac6b3f608b7fe56ee08a8b4699336e0b&vid=d2b49480ac6b3f608b7fe56ee08a8b4699336e0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR