2-Naphthalenesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Naphthalene-2-sulphonic acid, β-Naphthalenesulphonic acid

B0258-25G B0258-500G
TCB0258-025GEA 29.95 USD
TCB0258-025G TCB0258-500G
2-Naphthalenesulfonic acid ≥98.0% (by HPLC, titration analysis)
2-Naphthalenesulphonic acid

CAS Number: 76530-12-6
MDL Number: MFCD00004089
Molecular Formula: C10H8O3S
Molecular Weight: 208.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₈O₃S
MW: 208.24 g/mol
Boiling Pt: ∼350 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1.441 g/cm³ (25 °C)
MDL Number: MFCD00149271
CAS Number: 120-18-3
EINECS: 204-375-3
Merck Index: 12,06464
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ebca30e13a6286b6ab009f334de77b0a84b484d&vid=6ebca30e13a6286b6ab009f334de77b0a84b484d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR