5-Aminolevulinic acid hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
A0325-1G
A0325-100MG
TCA0325-001GEA
159.5
USD
TCA0325-001G
TCA0325-100MG
5-Aminolevulinic acid hydrochloride ≥98.0% (by titrimetric analysis)
5-Aminolevulinic acid hydrochloride
CAS Number: 5451-09-2
MDL Number: MFCD00012869
Molecular Formula: C5H9NO3
Molecular Weight: 167.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C5H10ClNO3 MW: 167.59 g/mol |
MDL Number:
MFCD00012869 CAS Number: 5451-09-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...