Benzoyl-β-alanine ≥98.0%

Supplier: TCI America

Synonyms: Betamipron, 3-Benzamidopropanoic acid

B0099-5G B0099-25G
TCB0099-5GEA 48 USD
TCB0099-5G TCB0099-025G
Benzoyl-β-alanine ≥98.0%
Benzoyl-beta-alanine

CAS Number: 3440-28-6
MDL Number: MFCD00037293
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 132


Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: Betamipron

Formula: C₁₀H₁₁NO₃
Melting Pt: 132 °C
MDL Number: MFCD00037293
CAS Number: 3440-28-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89fba5edb3e406c2e56e4f6496578be35ba5f6f5&vid=89fba5edb3e406c2e56e4f6496578be35ba5f6f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR