DL-3-Aminoisobutyric acid hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-Amino-2-methylpropanoic acid, BAIBA
A0324-5G
A0324-1G
TCA0324-5GEA
286
USD
TCA0324-5G
TCA0324-1G
DL-3-Aminoisobutyric acid hydrate ≥98.0% (by titrimetric analysis)
DL-3-Aminoisobutyric acid hydrate
CAS Number: 144-90-1
MDL Number: MFCD00008145
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method:
Formula:
C₄H₉NO₂·H₂O MW: 103.12 g/mol Melting Pt: 174 °C |
MDL Number:
MFCD00008145 CAS Number: 144-90-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...